Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Phosphotyrosine Mouse Monoclonal Antibody [clone: PY20]
Anti-Phosphotyrosine Mouse Monoclonal Antibody [clone: PY20]
Catalog # ICNA08691371
Supplier:  MP Biomedicals
Anti-Phosphotyrosine Mouse Monoclonal Antibody [clone: PY20]
Catalog # ICNA08691371
Supplier:  MP Biomedicals

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphotyrosin
  • Clonality:
    Monoclonal
  • Clone:
    PY20
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Storage temperature:
    0 to +5 °C
  • Concentration:
    1,0 mg/ml
  • Immunogen:
    Phosphotyrosine conjugated to carrier protein (PY-KLH)
  • Tested applications:
    Immunoblot
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

A large number of oncogene protein kinases and growth factor receptors will typically phosphorylate tyrosine residues. The PY20 and other phosphotyrosine antibodies have been effective in the characterization of tyrosine kinase substrates involved in oncogenesis, growth regulation and cell differentiation.

  • Working dilution: 1:500 for Immunoprecipitation, Immunohistochemistry, Immunocytochemistry
  • Working dilution: 1:5000 for Western Blot
  • Working dilution: 1:2000 for ELISA

Antibody is packaged in quantities of 100 µg at a concentration of 1 mg/ml in 20 mM sodium phosphate, 150 mM sodium chloride, pH 7,5. It is a highly purified immunoglobulin fraction.

It reacts specifically and strongly to phosphotyrosine containing proteins. It will not recognize phosphothreonine or phosphoserine containing proteins. This antibody is not species specific. It is known to be inhibited by divalent cations (>1 mM) and high salt concentrations (>0,2 M).

Type: Primary
Antigen: Phosphotyrosine
Clonality: Monoclonal
Clone: PY20
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: