Order Entry
Northern Ireland
ContactUsLinkComponent
Fish oil, from menhaden, clear yellow liquid, refined
Fish oil, from menhaden, clear yellow liquid, refined
Catalog # ICNA0296012001
Supplier:  MP Biomedicals
CAS Number:  
Fish oil, from menhaden, clear yellow liquid, refined
Catalog # ICNA0296012001
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    3,79 L
  • Seq:
    5134

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Typical Fatty Acid Composition (% wt.) for C20:5:
    14 ± 3%

Specification Test Results

About this item

 :
Storage Temperature:Freezer
CAS Number:8002-50-4
MDL Number:MFCD00131085