Potassium acetate ≥99.0%, white crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay:≥99,0%
- Heavy Metals:<5 ppm
- Insoluble Matter:<0,005%
- pH:6,5 - 9,0 (5% aq soln)
- Chloride:<0,003%
- Phosphate:<0,001%
- Sulphate:<0,002%
- Iron:<5 ppm
- Calcium:<0,005%
- Magnesium:<0,002%
- Sodium:<0,03%
Specification Test Results
Frequently Bought Together