L(+)-Lysine monohydrate, powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:39665-12-8
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MDL Number:MFCD00151035
:164.2 g/mol
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH (1% aq soln) :9.9 - 10.3
- Loss on Drying:≤11.8%
- Optical Rotation (c=8, 6 N HCl):+23.5° to +26.5°
Specification Test Results
Frequently Bought Together