Order Entry
Northern Ireland
ContactUsLinkComponent
Spectinomycin sulphate ≥98%, powder
Spectinomycin sulphate ≥98%, powder
Catalog # ICNA0215899302
Supplier:  MP Biomedicals
CAS Number:  
Spectinomycin sulphate ≥98%, powder
Catalog # ICNA0215899302
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :430.43 g/mol
CAS Number:23312-56-3
Formula:C₁₄H₂₄N₂O₇·H₂SO₄
MDL Number:MFCD00270185