Strontium chloride hexahydrate, crystalline powder ACS
Catalog # ICNA0215258390
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Synonyms:Strontium Dichloride Hexahydrate
- Form:White, Hard Packed, Crystalline Powder
- Storage:Store at Room Temperature (15 - 30 °C)
- Assay:99,0 - 103,0%
- Heavy Metals:NMT 0,0005% ; Iron NMT 0,0005%
- Insoluble Matter:NMT 0,005%
- pH:5,0 - 7,0 (5% sol)
- Sulfate NMT:0,001%
- Barium NMT:0,05%
- calcium NMT:0,05%
- Magnesium NMT:0,0002%
Specification Test Results
About this item
:
:266.62 g/mol
CAS Number:10025-70-4
Formula:SrCl₂·6H₂O
MDL Number:MFCD00149865
Boiling Pt:100
Melting Pt:61
Merck Index:13,08920