ContactUsLinkComponent
2,6-Dichlorophenolindophenol sodium salt, dark green powder indicator
2,6-Dichlorophenolindophenol sodium salt, dark green powder indicator
Supplier:  MP Biomedicals
Certificates
2,6-Dichlorophenolindophenol sodium salt, dark green powder indicator
Supplier:  MP Biomedicals
Specification Test Results
  • Identity Test:
    Passes
  • UV/VIS: λ max (water):
    604 ± 5 nm
  • Moisture:
    ≤12%