Esculin hydrate ≥97.5%, white powder
Catalog # ICNA0210165505
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:≥97.5%
- FTIR:Conforms to Standard
- UV/Vis: λmax (ethanol):334 - 338 nm
- pH (0,1% aq soln):4,6 - 5,2
- Loss on Drying:≤8%
- Optical Rotation:-84 ° to -87 °(c=2, 50% Dioxane)
- Solubility (0.1% aq soln):Clear and colourless within 15 minute
Specification Test Results
About this item
:
:340.29 g/mol (anhy)
Storage Temperature:Ambient
CAS Number:531-75-9
:MFCD00006879
Formula:C₁₅H₁₆O₉·xH₂O
MDL Number:MFCD00149492