Dextran, M.W. 35000 - 50000, powder
:9004-54-0
:
:(C₆H₁₀O₅)n
:MFCD00130935
:Ambient
- Identity Test:Passes
- Molecular Weight:35000 - 50000
- Specific Rotation:+192° to +204°
- Loss on Drying:≤5.0%
- pH:5.0 - 7.0
- Ash:Report result
Frequently Bought Together