
L(+)-Cysteine, white powder
:52-90-4
:
:HSCH₂CH(NH₂)CO₂H
:MFCD00064306
:121.16 g/mol
:200-158-2
:GHS07
:Ambient
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:213 - 235°C (dec)
- Optical Rotation:+7 ° to +13 ° (c=2, 1N HCl)
- pH:4 - 8 (5% aq soln)
- Refractive Index:8.8 ° (C=8, 1mol/L HCl) (Lit.)
Frequently Bought Together