Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:96
- Primary antibody reactivity:Mouse
- Target protein:TNFRSF17
- Description:Mouse TNFRSF17 ELISA Kit
- Detection range:31,2 - 2000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Mouse TNFRSF17 ELISA Kit for the quantitative detection of mouse tumour necrosis factor receptor superfamily member 17 (TNFRSF17, BCMA) in cell culture supernates, serum and plasma (heparin, EDTA).
TNFRSF17 is a protein that in humans is encoded by the TNFRSF17 gene. The protein encoded by this gene is a member of the TNF-receptor superfamily and is mapped to 16p13.13. This receptor is preferentially expressed in mature B lymphocytes, and may be important for B cell development and autoimmune response. This receptor has been shown to specifically bind to the tumor necrosis factor (ligand) superfamily, member 13b (TNFSF13B/TALL-1/BAFF), and to lead to NF-kappaB and MAPK8/JNK activation. This receptor also binds to various TRAF family members, and thus may transduce signals for cell survival and proliferation.