Order Entry
Northern Ireland
ContactUsLinkComponent
Human TNFSF13 ELISA kit
Human TNFSF13 ELISA kit
Catalog # BIRBORB96804-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human TNFSF13 ELISA kit
Catalog # BIRBORB96804-96
Supplier:  Biorbyt
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    TNFSF13
  • Description:
    Human TNFSF13 ELISA Kit
  • Specificity:
    Natural and recombinant human TNFSF13
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human TNFSF13 ELISA Kit is a sandwich ELISA kit for the quantitative detection of human TNFSF13.

  • No detectable cross-reactivity with other relevant proteins

Tumour necrosis factor ligand superfamily member 13 (TNFSF13) also known as a proliferation-inducing ligand (APRIL) is a protein that in humans is encoded by the TNFSF13 gene. TNFSF13 has also been designated CD256cluster of differentiation 256. The protein encoded by this gene is a member of the tumor necrosis factor ligand TNF ligand family. This protein is a ligand for TNFRSF17 or BCMA, a member of the TNF receptor family. This protein and its receptor are both found to be important for B cell development. In vivo experiments suggest an important role for APRIL in the long-term survival of plasma cells in the bone marrow. Mice deficient in April showed a reduced ability to support plasma cell survival In vitro experiments suggested that this protein may be able to induce apoptosis through its interaction with other TNF receptor family proteins such asTNFRSF6 or FAS and TNFRSF14 or HVEM. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. The TNFSF13 gene lies 878 bp downstream of the TNFSF12 gene on chromosome 17p13.1.