L(+)-Asparagine, anhydrous ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:70-47-3
:
Formula:H₂NCOCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064401
:132.12 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Loss on Drying:≤1,0%
- Optical Rotation (c=10, 1N HCl):+25° to +36°
Specification Test Results
Frequently Bought Together