Order Entry
Northern Ireland
ContactUsLinkComponent
Human HBEGF ELISA kit
Human HBEGF ELISA kit
Catalog # BIRBORB96788-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human HBEGF ELISA kit
Catalog # BIRBORB96788-96
Supplier:  Biorbyt
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Human
  • Description:
    Human HBEGF ELISA Kit
  • Specificity:
    Natural and recombinant human HBEGF
  • Detection range:
    15,6 - 1000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human HBEGF ELISA Kit is a sandwich ELISA kit for the quantitative detection of human heparin-binding EGF-like growth factor (HBEGF).

  • No detectable cross-reactivity with other relevant proteins

HBEGF, also known as HEGFL or DTR, is a member of the EGF family of proteins that in humans is encoded by the HBEGF gene. The HBEGF gene is assigned to chromosome 5, thus confirming the assignment of the gene on the basis of its role in relation to diphtheria toxin susceptibility. HBEGF is an 87 amino acid glycoprotein which displays highly regulated gene expression. It has been shown to play a role in wound healing, cardiac hypertrophy and heart development and function. HBEGF binding and activation of EGF receptors plays a critical role during cardiac valve tissue development and the maintenance of normal heart function in adults.