Urea, white prills, Reagent Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Boiling Pt:135 °C (275 °F) (dec., lit.)
Melting Pt:130…140 °C
Density:1,335 g/cm³ (20 °C) (lit.)
:60.06 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- OD260 nm (7 M aq soln):≤0.2
- OD280 (7M aq soln):≤0.2
- pH (7 M aq soln):7.5-9.5
Specification Test Results
Frequently Bought Together