Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GPI Rabbit Polyclonal Antibody
Anti-GPI Rabbit Polyclonal Antibody
Catalog # BSBTPB10069
Supplier:  Boster Bio
undefined
Anti-GPI Rabbit Polyclonal Antibody
Catalog # BSBTPB10069
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    GPI
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    14751
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Glucose-6-phosphate isomerase(GPI) detection. Tested with WB in Mouse.

Type: Primary
Antigen: GPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse