Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GPI Rabbit Polyclonal Antibody
Anti-GPI Rabbit Polyclonal Antibody
Catalog # BSBTPB10068
Supplier:  Boster Bio
undefined
Anti-GPI Rabbit Polyclonal Antibody
Catalog # BSBTPB10068
Supplier:  Boster Bio
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucose-6-phosphate isomerase
  • Antigen symbol:
    GPI
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    2821
  • Antigen synonyms:
    PGI|GNPI|SA36|AMF|PHI|NLK|SA-36
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Glucose-6-phosphate isomerase(GPI) detection. Tested with WB in Human.

Type: Primary
Antigen: GPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human