Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-FGF19 Rabbit Polyclonal Antibody
Anti-FGF19 Rabbit Polyclonal Antibody
Catalog # BSBTPB10061
Supplier:  Boster Bio
undefined
Anti-FGF19 Rabbit Polyclonal Antibody
Catalog # BSBTPB10061
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor 19
  • Antigen symbol:
    FGF19
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9965
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Fibroblast growth factor 19(FGF19) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: FGF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human