Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen symbol:CD21
- Clonality:Monoclonal
- Clone:TP1/31
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:914
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against human CD2.
Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: TP1/31
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human