Guard columns
Supplier: MACHEREY-NAGEL
Certificates
About this item
UNIVERSAL RP guard columns - to connect with the Column Protection System - are suitable for all HPLC columns under RP conditions.
- Economical protection for analytical main column
- Significant increase in column lifetime
Frequently Bought Together