ContactUsLinkComponent
Specific gravity hydrometers
Specific gravity hydrometers
Catalog # 10/903/0
Supplier:  S BRANNAN AND SONS
undefined
Specific gravity hydrometers
Catalog # 10/903/0
Supplier:  S BRANNAN AND SONS
Specifications
  • Length:
    190 mm
  • Seq:
    03
  • Division:
    0,002 °SG
  • Pk:
    1
  • Range:
    0,80 – 0,85 °SG
Specifications
About this item
Hydrometers for low surface tension.