ContactUsLinkComponent
Potassium bromide die set
Catalog # PERC51001144
Supplier:  PerkinElmer
Potassium bromide die set
Catalog # PERC51001144
Supplier:  PerkinElmer
Specifications
  • Description:
    Potassium bromide pellets die, Ø = 13 mm
  • Pk:
    1
Specifications
About this item
The evacuable potassium bromide (KBr) die is designed to produce uniform KBr pellets.
Suitable for FT-IR spectrometry of solid samples.