Order Entry
Northern Ireland
ContactUsLinkComponent
Clarithromycin 97%
Irritant
Clarithromycin 97%
Catalog # ACRO455220010
CAS Number:  
undefined
Irritant
Clarithromycin 97%
Catalog # ACRO455220010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    222 to 231 °C
  • Water:
    ≤2 %
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    -87° to -97° (20 °C, 589 nm) (c=1,CHCl3)

Specification Test Results

About this item

 :
 :747.96 g/mol
Melting Pt:222…231 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:81103-11-9
Formula:C₃₈H₆₉NO₁₃
MDL Number:MFCD00865140