Order Entry
Northern Ireland
ContactUsLinkComponent
 
4,4'-Dibromo-2,2'-bipyridyl 98%
 
undefined
4,4'-Dibromo-2,2'-bipyridyl 98%

 

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    140 to 145 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together