3-Buten-1-amine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2524-49-4
:
Formula:C₄H₉N
MDL Number:MFCD03425859
:71.12 g/mol
Boiling Pt:83 °C (760 mmHg)
Flash Pt:–5 °C
Storage Temperature:Ambient
UN:2733
ADR:3,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4215 to 1.4275 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together