Order Entry
Northern Ireland
ContactUsLinkComponent
Cadmium acetate, anhydrous 98%, pure
Irritant
Health
Environment
Cadmium acetate, anhydrous 98%, pure
Catalog # ACRO454002500
CAS Number:  
undefined
Irritant
Health
Environment
Cadmium acetate, anhydrous 98%, pure
Catalog # ACRO454002500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Complexometric:
    ≥97.5 %
  • Loss on drying:
    ≤0.5 %

Specification Test Results

About this item

 :
 :230.5 g/mol
Melting Pt:255…273 °C
Density:2,34 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2570
ADR:6.1,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:543-90-8
Formula:(H₃CCOO)₂Cd
MDL Number:MFCD00012449