
4-Chloro-3-iodobenzoic acid 97%
Catalog # ACRO452710050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:282.46 g/mol
Melting Pt:218…219,5 °C
:GHS07
Storage Temperature:Ambient
CAS Number:42860-04-8
Formula:C₇H₄ClIO₂
MDL Number:MFCD00079732