
2,4-Difluoro-3-nitrobenzonitrile 97%
Catalog # ACRO451420010
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
:
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS06
CAS Number:1186194-75-1
Formula:C₇H₂F₂N₂O₂
MDL Number:MFCD11226536