Order Entry
Northern Ireland
ContactUsLinkComponent
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98%
Irritant
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98%
Certificates
undefined
Irritant
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98%
Certificates

Specification Test Results

  • Appearance:
    White Crystalline solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -275° to -295° (20°C, 589 nm) (c=1, toluene)

Specification Test Results

Related Information

Frequently Bought Together