Order Entry
Northern Ireland
ContactUsLinkComponent
Mercury(I) nitrate dihydrate ACS (9th edition)
Toxic
Health
Environment
Mercury(I) nitrate dihydrate ACS (9th edition)
Certificates
undefined
Toxic
Health
Environment
Mercury(I) nitrate dihydrate ACS (9th edition)

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Titration Iodometric:
    ≥97.0 %
  • Additional info:
    Conform ACS 9th edition
  • Chloride (Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.005 %
  • Iron (Fe):
    ≤0.001 %
  • Mercuric Mercury (as Hg):
    ≤0.5 %
  • Insoluble matter:
    ≤0.005 % (in dilute HNO3)
  • Residue after reduction:
    ≤0.01 %

Specification Test Results

Related Information

Frequently Bought Together