

2,3-Difluoro-4-(trifluoromethyl)pyridine 97%
Catalog # ACRO446780010
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:183.08 g/mol
Boiling Pt:82…84 °C (400Torr)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:1215034-77-7
Formula:C₆H₂F₅N
MDL Number:MFCD16610846