Order Entry
Northern Ireland
ContactUsLinkComponent
2-(Trifluoromethyl)-1H-indole 97%
Irritant
2-(Trifluoromethyl)-1H-indole 97%
Catalog # ACRO446560010
CAS Number:  
undefined
Irritant
2-(Trifluoromethyl)-1H-indole 97%
Catalog # ACRO446560010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow to light brown Powder or solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :185.15 g/mol
Melting Pt:107…109 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:51310-54-4
Formula:C₉H₆F₃N
MDL Number:MFCD04972104