tri-Calcium phosphate 96%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-87-4
:MFCD00867081
Formula:Ca₃(PO₄)₂
MDL Number:MFCD00015984
:310.18 g/mol
Melting Pt:1670 °C
Density:3,14 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:≥95 %
Specification Test Results
Frequently Bought Together