Order Entry
Northern Ireland
ContactUsLinkComponent
Magnesium bromide hexahydrate 99%, extra pure
Irritant
Magnesium bromide hexahydrate 99%, extra pure
Certificates
undefined
Irritant
Magnesium bromide hexahydrate 99%, extra pure

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Complexometric:
    ≥98.5 %
  • pH:
    6 to 8 (5 % at 20°C)

Specification Test Results

Related Information

Frequently Bought Together