Phenylboronic acid pinacol ester 97%
Catalog # ACRO445220250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to white Low melting solid or liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:204.08 g/mol
Boiling Pt:110 °C (12Torr)
Melting Pt:27…31 °C
Flash Pt:107 °C
:GHS07
Storage Temperature:Ambient
CAS Number:24388-23-6
Formula:C₁₂H₁₇BO₂
MDL Number:MFCD00966985