Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
3,4-Diaminophenylboronic acid pinacol ester 97%
  ACRO444760010
 :  
undefined
 
3,4-Diaminophenylboronic acid pinacol ester 97%
  ACRO444760010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Tan to brown Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 100 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

 

 

 :
 :95…97 °C
 :234.106 g/mol
 :GHS07
 :Refrigerator
 :851883-08-4
 :C₁₂H₁₉BN₂O₂
 :MFCD09027073