ContactUsLinkComponent
D-Phenylalanine methyl ester hydrochloride 98%
D-Phenylalanine methyl ester hydrochloride 98%
Certificates
undefined
D-Phenylalanine methyl ester hydrochloride 98%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    -32° to -38° (20°C, 589 nm) (c=1 in EtOH)