N-Boc-L-serine, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate
CAS Number:3262-72-4
:
Formula:C₈H₁₅NO₅
MDL Number:MFCD00150501
:205.21 g/mol
Melting Pt:84…89 °C
Flash Pt:186 °C
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:≤1 %
- HPLC:≥98.5 %
- Specific optical rotation:-6.5° to -8.5° (20°C, 589 nm) (c=2 in water)
Specification Test Results
Frequently Bought Together