



2-Benzyloxyphenylmagnesium bromide 1 M in THF, AcroSeal®
Catalog # 440320500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear to hazy yellow to brown Solution
- Titration Grignard:0.95 to 1.10 M
Specification Test Results
About this item
:
:287.44 g/mol
Density:1,047 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:328000-16-4
Formula:C₁₃H₁₁BrMgO
MDL Number:MFCD08705262