
Methyl-2-amino-4-chlorobenzoate 95%
Catalog # ACRO439231000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:White to light brown Powder
- Infrared spectrum:Conforms
- Melting point:65 to 71 °C
- HPLC:≥94.0 %
Specification Test Results
About this item
:
:185.61 g/mol
Boiling Pt:106 °C (8 mmHg)
Melting Pt:65…67 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5900-58-3
Formula:C₈H₈ClNO₂
MDL Number:MFCD00017568