ContactUsLinkComponent
Ethynylmagnesium bromide 0.5 M in THF, AcroSeal®
Flammable
Corrosive
Irritant
Health
Ethynylmagnesium bromide 0.5 M in THF, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Ethynylmagnesium bromide 0.5 M in THF, AcroSeal®
Specification Test Results
  • Appearance:
    Colorless to yellow to grey to brown Solution (May develop some turbidity or precipitate)
  • Titration Grignard:
    0.40 to 0.60 M