Order Entry
Northern Ireland
ContactUsLinkComponent
O-Phenylhydroxylamine hydrochloride 97%
Toxic
O-Phenylhydroxylamine hydrochloride 97%
Catalog # ACRO438810010
CAS Number:  
undefined
Toxic
O-Phenylhydroxylamine hydrochloride 97%
Catalog # ACRO438810010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to off-white to light brown Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :145.586 g/mol
Melting Pt:121…122 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:6092-80-4
Formula:C₆H₈ClNO
MDL Number:MFCD00043271