

FMOC isothiocyanate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:199915-38-3
:
Formula:C₁₆H₁₁NO₂S
MDL Number:MFCD01861169
Melting Pt:44…45 °C
:281.329 g/mol
:GHS05
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to yellow Low melting solid
- Infrared spectrum:Conforms
- Elemental Analysis:65.6 to 71.0 % (C)
Specification Test Results
Frequently Bought Together