Methyl tridecanoate 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Tridecylic acid methyl ester, Tridecanoic acid methyl ester
- CAS Number:1731-88-0
- Formula:C₁₄H₂₈O₂
- MDL Number:MFCD00008977
- Molecular Weight:228.38 g/mol
- Boiling Pt:130…132 °C (4Torr)
- Melting Pt:7 °C
- Density:0,86 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Tridecylic acid methyl ester, Tridecanoic acid methyl ester
- CAS Number:1731-88-0
- Formula:C₁₄H₂₈O₂
- MDL Number:MFCD00008977
- Molecular Weight:228.38 g/mol
- Boiling Pt:130…132 °C (4Torr)
- Melting Pt:7 °C
- Density:0,86 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Ambient
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



