

tetra-Chloroauric(III) acid hydrate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27988-77-8
:
Formula:HAuCl₄·H₂O
MDL Number:MFCD00149903
:357.8 g/mol
Melting Pt:30 °C
Density:1,169 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:3260
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Yellow to orange Crystals
- Assay:49 to 51 % (Au)
- Total trace metal impurities:≤10 ppm
Specification Test Results
Frequently Bought Together