Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium bromide 99.999% (trace metals basis)
Irritant
Potassium bromide 99.999% (trace metals basis)
Certificates
undefined
Irritant
Potassium bromide 99.999% (trace metals basis)

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Argentometric:
    33 % (K) Typical
  • Total trace metal impurities:
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together