


Triethyloxonium tetrafluoroborate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:368-39-8
:
Formula:C₆H₁₅BF₄O
MDL Number:MFCD00044423
:189.99 g/mol
Melting Pt:84…89 °C
UN:2925
ADR:4.1,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Stabilizer:3 to 5 % ether
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together