
Diethyl carbonate, anhydrous 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105-58-8
:
Formula:C₅H₁₀O₃
MDL Number:MFCD00009107
:118.13 g/mol
Boiling Pt:126…128 °C (760 mmHg)
Melting Pt:–43 °C
Density:0,975 g/cm³
Flash Pt:33 °C
Storage Temperature:Ambient
UN:2366
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3830 to 1.3850 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.5 %
Specification Test Results
Related Information
Frequently Bought Together