ContactUsLinkComponent
p-Toluenesulphinic acid sodium salt, anhydrous 97%
p-Toluenesulphinic acid sodium salt, anhydrous 97%
Certificates
undefined
p-Toluenesulphinic acid sodium salt, anhydrous 97%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤3.00 %
  • HPLC:
    ≥96.0 %