Order Entry
Northern Ireland
ContactUsLinkComponent
(R)-(-)-3-Hydroxytetrahydrofuran 98%
Irritant
(R)-(-)-3-Hydroxytetrahydrofuran 98%
Catalog # ACRO434070010
CAS Number:  
undefined
Irritant
(R)-(-)-3-Hydroxytetrahydrofuran 98%
Catalog # ACRO434070010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -15.0° to -21.0° (20°C, 589 nm) (c=2.4, methanol)

Specification Test Results

About this item

 :
 :88.11 g/mol
Boiling Pt:181 °C
Flash Pt:82 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:86087-24-3
Formula:C₄H₈O₂
MDL Number:MFCD00067101